Invitrogen
Manufacturer Code:PA570457
Catalog # PIPA570457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human HHIPL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARAR9245; hedgehog interacting protein-like 1; HHIP-like 1; HHIP-like protein 1
Gene Aliases: 1600002O04Rik; AK129450; HHIP2; HHIPL1; KIAA1822; mKIAA1822; RGD1311698; UNQ9245; UNQ9245/PRO34761
UniProt ID: (Human) Q96JK4, (Mouse) Q14DK5
Entrez Gene ID: (Dog) 612500, (Pig) 100625046, (Horse) 100051205, (Human) 84439, (Rabbit) 100354830, (Mouse) 214305, (Rat) 362781, (Guinea pig) 100729505, (Zebrafish) 100002738
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.