Invitrogen
Manufacturer Code:PA5113265
Catalog # PIPA5113265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Hedgehog-interacting protein (18-220AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hedgehog interacting protein Hedgehog-interacting protein Hedgehog-interacting protein-like HHIP Hhip1 HIP RGD1564108 UNQ5825/PRO19644; Hedgehog-interacting protein; Hedgehog-interacting protein-like; HHIP
Gene Aliases: HHIP; HIP; RGD1564108; UNQ5825/PRO19644
UniProt ID: (Human) Q96QV1
Entrez Gene ID: (Human) 64399, (Rat) 291936
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.