Novus Biologicals
Manufacturer Code:NBP270890JF646
Catalog # N270890J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3E6 |
Immunogen | Recombinant fragment expressed in E.coli corresponding to amino acids 100-270 of human HHEX. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: hematopoietically expressed homeobox HEXHMPH Homeobox protein HEX Homeobox protein PRH HOX11L-PEN PRHhematopoietically-expressed homeobox protein HHEX PRHXhomeobox hematopoietically expressed proline-rich homeodomain-containing transcription factor; Hematopoietically-expressed homeobox protein HHEX; Homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; proline-rich homeodomain-containing transcription factor
Gene Aliases: HEX; HHEX; HMPH; HOX11L-PEN; PRH; PRHX
UniProt ID: (Human) Q03014
Entrez Gene ID: (Human) 3087
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.