Abnova
Manufacturer Code:H00003087M13
Catalog # 89-022-483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B12 |
Immunogen | HHEX (NP_002720 133 a.a. ~ 237 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant HHEX.
For Research Use Only
Protein Aliases: Hematopoietically-expressed homeobox protein HHEX; HEX; HMPH; Homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; HOX11L-PEN; PRH; PRHX; proline-rich homeodomain-containing transcription factor
Gene Aliases: HEX; HHEX; HMPH; HOX11L-PEN; PRH; PRHX
UniProt ID: (Human) Q03014
Entrez Gene ID: (Human) 3087
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.