R&D Systems
Manufacturer Code:MAB83771100
Catalog # MAB83771100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2018B |
Immunogen | <em>E. coli</em>-derived recombinant human HHEX Thr111-Gly270 Accession # Q03014 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: hematopoietically expressed homeobox Hex HEXHMPH HMPH Homeobox protein HEX Homeobox protein PRH HOX11L-PEN PRH PRHhematopoietically-expressed homeobox protein HHEX PRHX PRHXhomeobox hematopoietically expressed proline-rich homeodomain-containing transcription factor; Hematopoietically-expressed homeobox protein HHEX; Homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; proline-rich homeodomain-containing transcription factor
Gene Aliases: HEX; HHEX; HMPH; HOX11L-PEN; PRH; PRHX
UniProt ID: (Human) Q03014
Entrez Gene ID: (Human) 3087
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.