Abnova
Manufacturer Code:H00057467M07A
Catalog # 89-107-184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B7 |
Immunogen | HHATL (NP_065758.2157 a.a. &bsim 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HHATL.
For Research Use Only
Protein Aliases: C3orf3; Glycerol uptake/transporter homolog; GUP1; GUP1 glycerol uptake/transporter homolog; Hedgehog acyltransferase-like protein; KIAA1173; MBOAT3; membrane bound O-acyltransferase domain containing 3; MSTP002; OACT3; Protein-cysteine N-palmitoyltransferase HHAT-like protein
Gene Aliases: C3orf3; GUP1; HHATL; KIAA1173; MBOAT3; MSTP002; OACT3
UniProt ID: (Human) Q9HCP6
Entrez Gene ID: (Human) 57467
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.