Novus Biologicals
Manufacturer Code:NBP304745100UL
Catalog # NB096087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 160-245 of human HHATL (NP_065758.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C3orf3 chromosome 3 open reading frame 3 glycerol uptake/transporter homolog GUP1 hedgehog acyltransferase-like Hedgehog acyltransferase-like protein KIAA1173GUP1 glycerol uptake/transporter homolog (yeast) MBOAT3 membrane bound O-acyltransferase domain containing 3 protein-cysteine N-palmitoyltransferase HHAT-like protein; Glycerol uptake/transporter homolog; GUP1 glycerol uptake/transporter homolog; Hedgehog acyltransferase-like protein; membrane bound O-acyltransferase domain containing 3; Protein-cysteine N-palmitoyltransferase HHAT-like protein
Gene Aliases: C3orf3; GUP1; HHATL; KIAA1173; MBOAT3; MSTP002; OACT3
UniProt ID: (Human) Q9HCP6
Entrez Gene ID: (Human) 57467
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.