Invitrogen
Manufacturer Code:PA553425
Catalog # PIPA553425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HHAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hedgehog acyltransferase; MART-2; MART2 SKI1 Skn melanoma antigen recognized by T cells 2 melanoma antigen recognized by T-cells 2 protein-cysteine N-palmitoyltransferase HHAT skinny hedgehog protein 1; melanoma antigen recognized by T cells 2; Melanoma antigen recognized by T-cells 2; Protein-cysteine N-palmitoyltransferase HHAT; Skinny hedgehog protein 1
Gene Aliases: HHAT; MART2; SKI1; Skn
UniProt ID: (Human) Q5VTY9
Entrez Gene ID: (Human) 55733
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.