Invitrogen
Manufacturer Code:PA5106464
Catalog # PIPA5106464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MEKKK 4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430080K19Rik AU043147 AU045934 AW046177 epididymis secretory protein Li 31 FLH21957 FLJ10410 FLJ20373 FLJ90111 HEL-S-31 hepatocyte progenitor kinase-like/germinal center kinase-like kinase HGK HPK/GCK-like kinase HGK KIAA0687 MAP4K4 MAPK/ERK kinase kinase kinase 4 MEK kinase kinase 4 MEKKK 4 MEKKK4 mitogen activated protein kinase kinase kinase kinase 4 mitogen-activated protein kinase kinase kinase kinase 4 NCK interacting kinase nck-interacting kinase NIK Ste20 group protein kinase HGK; epididymis secretory protein Li 31; hepatocyte progenitor kinase-like/germinal center kinase-like kinase; HPK/GCK-like kinase HGK; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; MEKKK 4; mitogen activated protein kinase kinase kinase kinase 4; Mitogen-activated protein kinase kinase kinase kinase 4; NCK interacting kinase; Nck-interacting kinase; Ste20 group protein kinase HGK
Gene Aliases: 9430080K19Rik; AU043147; AU045934; AW046177; FLH21957; HEL-S-31; HGK; KIAA0687; MAP4K4; MEKKK4; NIK
UniProt ID: (Human) O95819, (Mouse) P97820
Entrez Gene ID: (Human) 9448, (Mouse) 26921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.