R&D Systems
Manufacturer Code:AF276SP
Catalog # AF276SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human HGF R/c-MET Glu25-Thr932 Accession # P08581 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AUTS9 c-Met cMET EC 2.7.10 EC 2.7.10.1 hepatocyte growth factor receptor HGF R HGF receptor HGF/SF receptor HGFR MET Met (c-Met) met proto-oncogene (hepatocyte growth factor receptor) met proto-oncogene tyrosine kinase oncogene MET Proto-oncogene c-Met RCCP2 Scatter factor receptor SF receptor Tyrosine-protein kinase Met; Hepatocyte growth factor receptor; HGF receptor; HGF/SF receptor; Proto-oncogene c-Met; Scatter factor receptor; SF receptor; Tyrosine-protein kinase Met
Gene Aliases: AUTS9; c-Met; DFNB97; HGFR; MET; RCCP2
UniProt ID: (Human) P08581
Entrez Gene ID: (Human) 4233
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.