Novus Biologicals
Manufacturer Code:NBP119182
Catalog # NBP119182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 43283 |
Immunogen | Human Hepatocyte Growth Factor/Scatter Factor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: deafness autosomal recessive 39 DFNB39 EC 3.4.21 EC 3.4.21.7 fibroblast-derived tumor cytotoxic factor F-TCF hepatocyte growth factor (hepapoietin A scatter factor) Hepatopoeitin-A HGFB HPTAhepatocyte growth factor lung fibroblast-derived mitogen Scatter factor SFhepatopoeitin-A; fibroblast-derived tumor cytotoxic factor; Hepatocyte growth factor; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoietin-A; lung fibroblast-derived mitogen; Scatter factor; SF
Gene Aliases: DFNB39; F-TCF; HGF; HGFB; HPTA; SF
UniProt ID: (Human) P14210
Entrez Gene ID: (Human) 3082
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.