R&D Systems
Manufacturer Code:MAB294500
Catalog # MAB294500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 24612 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human HGF Gln32-Ser728 Accession # P14210 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: deafness autosomal recessive 39 DFNB39 EC 3.4.21 EC 3.4.21.7 fibroblast-derived tumor cytotoxic factor F-TCF hepatocyte growth factor (hepapoietin A scatter factor) Hepatopoeitin-A Hepatopoietin A HGFB HPTA HPTAhepatocyte growth factor lung; fibroblast-derived tumor cytotoxic factor; Hepatocyte growth factor; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoietin-A; lung fibroblast-derived mitogen; Scatter factor; SF
Gene Aliases: DFNB39; F-TCF; HGF; HGFB; HPTA; SF
UniProt ID: (Human) P14210
Entrez Gene ID: (Human) 3082
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.