Invitrogen
Manufacturer Code:PA537364
Catalog # PIPA537364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human HFE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hemochromatosis Hemochromatosis protein Hereditary hemochromatosis protein Hereditary hemochromatosis protein HLA H HFE 1 HFE HFE HFE1 HH High Fe HLA H HLA-H HLAH MGC:150812 MGC10379 MGC103790 MHC class I like protein HFE MVCD7 TFQTL2; Hereditary hemochromatosis protein; hereditary hemochromatosis protein HLA-H; high Fe; HLA-H; MHC class I-like protein HFE
Gene Aliases: HFE; HFE1; HH; HLA-H; HLAH; MVCD7; TFQTL2
UniProt ID: (Human) Q30201
Entrez Gene ID: (Human) 3077
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.