Novus Biologicals
Manufacturer Code:NBP299531FR
Catalog # NB061387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse HFE. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ221C16.10.1 hemochromatosis hereditary hemochromatosis protein hereditary hemochromatosis protein HLA-H HH high Fe HLAH HLA-HHFE1 MGC103790 MHC class I-like protein HFE MVCD7; Hereditary hemochromatosis protein; hereditary hemochromatosis protein HLA-H; high Fe; HLA-H; MHC class I-like protein HFE
Gene Aliases: HFE; HFE1; HH; HLA-H; HLAH; MVCD7; TFQTL2
UniProt ID: (Human) Q30201
Entrez Gene ID: (Human) 3077
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.