Abnova
Manufacturer Code:PAB25214
Catalog # 89-126-885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to HFE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of HFE.
For Research Use Only
Protein Aliases: dJ221C16.10.1; Hereditary hemochromatosis protein; hereditary hemochromatosis protein HLA-H; HFE1; HH; high Fe; HLA-H; MGC103790; MHC class I-like protein HFE
Gene Aliases: HFE; HFE1; HH; HLA-H; HLAH; MVCD7; TFQTL2
UniProt ID: (Human) Q30201
Entrez Gene ID: (Human) 3077
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.