Abnova
Manufacturer Code:H00026508M22
Catalog # 89-002-621
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E12 |
Immunogen | HEYL (NP_055386 1 a.a. ~ 70 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HEYL.
For Research Use Only
Protein Aliases: bHLHb33; Class B basic helix-loop-helix protein 33; Hairy-related transcription factor 3; hairy/enhancer-of-split related with YRPW motif 3; Hairy/enhancer-of-split related with YRPW motif-like protein; hes-related family bHLH transcription factor with YRPW motif-like; HEY-like protein; hHeyL; hHRT3; HRT-3; HRT3; MGC12623
Gene Aliases: BHLHB33; HESR3; HEY3; HEYL; HRT3
UniProt ID: (Human) Q9NQ87
Entrez Gene ID: (Human) 26508
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.