Invitrogen
Manufacturer Code:PA535185
Catalog # PIPA535185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human HEYL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb33; Class B basic helix-loop-helix protein 33; Hairy-related transcription factor 3; hairy/enhancer-of-split related with YRPW motif 3; Hairy/enhancer-of-split related with YRPW motif-like protein; hes-related family bHLH transcription factor with YRPW motif-like; HEY-like protein; HEYL BHLHB33 HRT3 Hairy/enhancer-of-split related with YRPW motif-like protein Class B basic helix-loop-helix protein 33 Hairy-related transcription factor 3; hHeyL; hHRT3; HRT-3
Gene Aliases: BHLHB33; HESR3; HEY3; HEYL; HRT3
UniProt ID: (Human) Q9NQ87
Entrez Gene ID: (Human) 26508
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.