Invitrogen
Manufacturer Code:PA5116766
Catalog # PIPA5116766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 129-328 of human HEYL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb33; bHLHb33 Class B basic helix-loop-helix protein 33 hairy and enhance of split related 3 hairy and enhancer of split-related protein 3 hairy/enhancer-of-split related with YRPW motif 3 hairy/enhancer-of-split related with YRPW motif-like hairy/enhancer-of-split related with YRPW motif-like protein Hairy-related transcription factor 3 hes related family bHLH transcription factor with YRPW motif-like Hesr3 hes-related family bHLH transcription factor with YRPW motif-like HEY3 HEYL HEY-like protein hHeyL hHRT3 Hrt3 HRT-3 mHRT3; Class B basic helix-loop-helix protein 33; hairy and enhance of split related 3; Hairy and enhancer of split-related protein 3; Hairy-related transcription factor 3; hairy/enhancer-of-split related with YRPW motif 3; Hairy/enhancer-of-split related with YRPW motif-like protein; hes-related family bHLH transcription factor with YRPW motif-like; HEY-like protein; hHeyL; hHRT3; HRT-3; mHRT3
Gene Aliases: BHLHB33; HESR3; HEY3; HEYL; HRT3
UniProt ID: (Human) Q9NQ87, (Mouse) Q9DBX7
Entrez Gene ID: (Human) 26508, (Mouse) 56198
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.