Invitrogen
Manufacturer Code:PA572676
Catalog # PIPA572676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 224-260 amino acids from the C-terminal region of Zebrafish hey2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix transcription factor; CHF1 GRIDLOCK GRL HERP1 HESR2 HRT2 bHLHb32 HES-related repressor protein 1 HES-related repressor protein 2 HESR-2 HRT-2 cardiovascular basic helix-loop-helix factor 1 cardiovascular helix-loop-helix factor 1 class B basic helix-loop-helix protein 32 hCHF1 hHRT2 hairy and enhancer of split-related; gridlock; hairy/enhancer-of-split related with YRPW motif 2; Hairy/enhancer-of-split related with YRPW motif protein 2; Herp2; Protein gridlock
Gene Aliases: grl; hey2; zgc:136746
UniProt ID: (Zebrafish) Q9I9L0
Entrez Gene ID: (Zebrafish) 58146
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.