Invitrogen
Manufacturer Code:PA567365
Catalog # PIPA567365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human HEY2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb32; cardiovascular basic helix-loop-helix factor 1; Cardiovascular helix-loop-helix factor 1; CHF1 GRIDLOCK GRL HERP1 HESR2 HRT2 bHLHb32 HES-related repressor protein 1 HES-related repressor protein 2 HESR-2 HRT-2 cardiovascular basic helix-loop-helix factor 1 cardiovascular helix-loop-helix factor 1 class B basic helix-loop-helix protein 32 hCHF1 hHRT2 hairy and enhancer of split-related; Class B basic helix-loop-helix protein 32; Hairy and enhancer of split-related protein 2; Hairy-related transcription factor 2; hairy/enhancer-of-split related with YRPW motif 2; Hairy/enhancer-of-split related with YRPW motif protein 2; hCHF1; hes-related family bHLH transcription factor with YRPW motif 2; HES-related repressor protein 1; HES-related repressor protein 2; HESR-2; hHRT2; HRT-2; Protein gridlock homolog
Gene Aliases: BHLHB32; CHF1; GRIDLOCK; GRL; HERP; HERP1; HESR2; HEY2; HRT2
UniProt ID: (Human) Q9UBP5
Entrez Gene ID: (Human) 23493
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.