Novus Biologicals
Manufacturer Code:NBP188629
Catalog # NBP188629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:YLSSVEGLDSSDPLRVRLVSHLSTCATQREAAAMTSSMAHHHHPLHPHHWAAAFHHLPAALLQPN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: bHLHb32; BHLHB32 bHLHb32HES-related repressor protein 2 cardiovascular basic helix-loop-helix factor 1 Cardiovascular helix-loop-helix factor 1 CHF1 Class B basic helix-loop-helix protein 32 GRLGRIDLOCK Hairy and enhancer of split-related protein 2 hairy/enhancer-of-split related with YRPW motif 2 hairy/enhancer-of-split related with YRPW motif protein 2 Hairy-related transcription factor 2 hCHF1 HERP HERP1hHRT2 HESR2 HESR-2 HES-related repressor protein 1 HRT2 HRT-2 MGC10720 Protein gridlock homolog; cardiovascular basic helix-loop-helix factor 1; Cardiovascular helix-loop-helix factor 1; Class B basic helix-loop-helix protein 32; Hairy and enhancer of split-related protein 2; Hairy-related transcription factor 2; hairy/enhancer-of-split related with YRPW motif 2; Hairy/enhancer-of-split related with YRPW motif protein 2; hCHF1; hes-related family bHLH transcription factor with YRPW motif 2; HES-related repressor protein 1; HES-related repressor protein 2; HESR-2; hHRT2; HRT-2; Protein gridlock homolog
Gene Aliases: BHLHB32; CHF1; GRIDLOCK; GRL; HERP; HERP1; HESR2; HEY2; HRT2
UniProt ID: (Human) Q9UBP5
Entrez Gene ID: (Human) 23493
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.