Invitrogen
Manufacturer Code:PA542008
Catalog # PIPA542008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human HEY1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix protein OAF1; basic helix-loop-helix transcription factor; bHLH protein Hesr-1/Hey1; bHLHb31; cardiovascular basic helix-loop-helix factor 2; Cardiovascular helix-loop-helix factor 2; CHF-2; Class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy-related transcription factor 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif 1; Hairy/enhancer-of-split related with YRPW motif 1 CHF2 HERP2 HESR1 HRT-1 MGC1274 OAF1 BHLHb31; Hairy/enhancer-of-split related with YRPW motif protein 1; Herp1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; HESR-1; HRT-1; mHRT1
Gene Aliases: AI316788; AI414254; BC8; BHLHB31; CHF2; HERP1; HERP2; hesr-1; HESR1; HEY1; hHRT1; HRT-1; HRT1; id:ibd1292; OAF1; zgc:110572
UniProt ID: (Dog) Q9TSZ2, (Bovine) Q2KIN4, (Human) Q9Y5J3, (Zebrafish) Q8AXV6
Entrez Gene ID: (Dog) 403420, (Horse) 100058549, (Bovine) 408005, (Sheep) 101111557, (Rabbit) 100344717, (Human) 23462, (Mouse) 15213, (Rat) 155437, (Guinea pig) 100734127, (Zebrafish) 58008
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.