Novus Biologicals
Manufacturer Code:NBP227152SS
Catalog # NBP227152SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 50-100 of mouse Hey1 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix protein OAF1; basic helix-loop-helix protein OAF1 BHLHb31 Cardiovascular helix-loop-helix factor 2 CHF-2 CHF2hHRT1 Class B basic helix-loop-helix protein 31 Hairy and enhancer of split-related protein 1 hairy/enhancer-of-split related with YRPW motif 1 hairy/enhancer-of-split related with YRPW motif protein 1 Hairy-related transcription factor 1 HERP2HES-related repressor protein 1 HESR-1 HESR1MGC1274 HES-related repressor protein 2 HRT1 HRT-1OAF1; bHLHb31; Cardiovascular helix-loop-helix factor 2; CHF-2; Class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy-related transcription factor 1; hairy/enhancer-of-split related with YRPW motif 1; Hairy/enhancer-of-split related with YRPW motif protein 1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; HESR-1; HRT-1
Gene Aliases: BHLHB31; CHF2; HERP2; HESR1; HEY1; hHRT1; HRT-1; HRT1; OAF1
UniProt ID: (Human) Q9Y5J3
Entrez Gene ID: (Human) 23462
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.