Novus Biologicals
Manufacturer Code:NBP272312JF549
Catalog # N272312J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B3 |
Immunogen | Full length recombinant protein of human HEXO (NP_699163) produced in HEK293T cells. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 3&prime exoribonuclease 3&prime-5&prime exonuclease ERI1 3&primeEXO 3&primeHEXO EC 3.1 enhanced RNAi three prime mRNA exonuclease homolog 1 Eri-1 homolog exoribonuclease 1 HEXO histone mRNA 3&prime end-specific exonuclease Histone mRNA 3&prime-end-specific exoribonuclease Histone mRNA 3&prime-exonuclease 1 MGC35395 Protein 3&primehExo THEX1 three prime histone mRNA exonuclease 13&prime-5&prime exoribonuclease 1 three prime mRNA exonuclease 1; 3' exoribonuclease; 3'-5' exonuclease ERI1; 3'-5' exoribonuclease 1; enhanced RNAi three prime mRNA exonuclease homolog 1; Eri-1 homolog; HEXO; histone mRNA 3' end-specific exonuclease; Histone mRNA 3'-end-specific exoribonuclease; Histone mRNA 3'-exonuclease 1; Protein 3'hExo; three prime histone mRNA exonuclease 1; three prime mRNA exonuclease 1
Gene Aliases: 3'EXO; 3'HEXO; ERI1; HEXO; THEX1
UniProt ID: (Human) Q8IV48
Entrez Gene ID: (Human) 90459
Molecular Function: RNA binding protein esterase exoribonuclease hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.