Invitrogen
Manufacturer Code:PA578336
Catalog # PIPA578336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human HEXIM1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac lineage protein 1; CLP1 EDG1 FLJ13562 HIS1 MAQ1 HEXIM1 estrogen down-regulated gene 1 protein menage a quatre 1 menage a quatre protein 1 hexamethylene-bis-acetamide-inducible transcript 1 hexamthylene bis-acetamide inducible 1 HMBA-inducible cardiac lineage protein 1 hexamethylene bis-acetamide-inducible protein 1 hexamethylene bisacetamide-inducible protein protein HEXIM1 hexamethylene bis-acetamide inducible 1; Estrogen down-regulated gene 1 protein; hexamethylene bis-acetamide inducible 1; Hexamethylene bis-acetamide-inducible protein 1; hexamethylene bisacetamide-inducible protein; hexamethylene-bis-acetamide-inducible transcript 1; hexamthylene bis-acetamide inducible 1; HMBA-inducible; menage a quatre 1; Menage a quatre protein 1; Protein HEXIM1
Gene Aliases: CLP1; EDG1; HEXIM1; HIS1; MAQ1
UniProt ID: (Human) O94992
Entrez Gene ID: (Human) 10614
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.