Invitrogen
Manufacturer Code:PA538554
Catalog # PIPA538554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human HES7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac lineage protein 1; Estrogen down-regulated gene 1 protein; hexamethylene bis-acetamide inducible 1; Hexamethylene bis-acetamide-inducible protein 1; hexamethylene bisacetamide-inducible protein; hexamethylene-bis-acetamide-inducible transcript 1; hexamthylene bis-acetamide inducible 1; HEXIM1 hexamethylene bis-acetamide inducible 1 HGNC:24953 CLP1 HIS1 MAQ1 HMBA-inducible hexamethylene bisacetamide-inducible protein hexamethylene-bis-acetamide-inducible transcript 1 hexamthylene bis-acetamide inducible 1 menage a quatre 1 EDG1 FLJ13562; HMBA-inducible; menage a quatre 1; Menage a quatre protein 1; Protein HEXIM1
Gene Aliases: 7330426E13Rik; CLP-1; CLP1; EDG1; HEXIM1; HIS1; MAQ1
UniProt ID: (Human) O94992, (Mouse) Q8R409, (Rat) Q5M9G1
Entrez Gene ID: (Human) 10614, (Mouse) 192231, (Rat) 498008
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.