Invitrogen
Manufacturer Code:PA5116395
Catalog # PIPA5116395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 135-166 amino acids from human HEXDC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC069960 Beta-hexosaminidase D beta-N-acetylhexosaminidase Hexd Hexdc hexosaminidase (glycosyl hydrolase family 20 catalytic domain) containing hexosaminidase D hexosaminidase D cytosolic hexosaminidase domain-containing protein N-acetyl-beta-galactosaminidase; Beta-hexosaminidase D; Beta-N-acetylhexosaminidase; hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing; Hexosaminidase D; hexosaminidase D, cytosolic; Hexosaminidase domain-containing protein; N-acetyl-beta-galactosaminidase
Gene Aliases: HEXD; HEXDC
UniProt ID: (Human) Q8WVB3
Entrez Gene ID: (Human) 284004
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.