Novus Biologicals
Manufacturer Code:NBP183573
Catalog # NBP183573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SARCLPPRCHPPALAGTLLRTPEGRAHARGLLLEAGGALHCQMAWAIRAHVGVVPSGPAVSCPHSVPEGPGQPLGERLENTEGSSTGRPAL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-hexosaminidase D; Beta-hexosaminidase D Beta-N-acetylhexosaminidase EC 3.2.1.52 FLJ23825 hexosaminidase (glycosyl hydrolase family 20 catalytic domain) containing hexosaminidase D Hexosaminidase domain-containing protein N-acetyl-beta-galactosaminidase; Beta-N-acetylhexosaminidase; hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing; Hexosaminidase D; hexosaminidase D, cytosolic; Hexosaminidase domain-containing protein; N-acetyl-beta-galactosaminidase
Gene Aliases: HEXD; HEXDC
UniProt ID: (Human) Q8WVB3
Entrez Gene ID: (Human) 284004
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.