Invitrogen
Manufacturer Code:PA538571
Catalog # PIPA538571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human HEXB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta Beta-N-acetylhexosaminidase subunit beta Hexosaminidase subunit B Cervical cancer proto-oncogene 7 protein HCC-7 N-acetyl-beta-glucosaminidase subunit beta HEXB HCC7; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; epididymis luminal protein 248; epididymis secretory protein Li 111; HCC-7; hexosaminidase B (beta polypeptide); Hexosaminidase subunit B; N-acetyl-beta-glucosaminidase subunit beta
Gene Aliases: ENC-1AS; HCC7; HEL-248; HEL-S-111; HEXB
UniProt ID: (Human) P07686
Entrez Gene ID: (Human) 3074
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.