Invitrogen
Manufacturer Code:PA5101082
Catalog # PIPA5101082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HEXB corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta Beta-hexosaminidase subunit beta chain A Beta-hexosaminidase subunit beta chain B Beta-N-acetylhexosaminidase subunit beta cervical cancer proto-oncogene 7 protein ENC-1AS epididymis luminal protein 248 epididymis secretory protein Li 111 HCC7 HCC-7 HEL-248 HEL-S-111 HEXB hexosaminidase B hexosaminidase B (beta polypeptide) hexosaminidase subunit B hexosaminidase subunit beta N-acetyl-beta-glucosaminidase subunit beta; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; epididymis luminal protein 248; epididymis secretory protein Li 111; HCC-7; hexosaminidase B (beta polypeptide); Hexosaminidase subunit B; N-acetyl-beta-glucosaminidase subunit beta
Gene Aliases: ENC-1AS; HCC7; HEL-248; HEL-S-111; HEXB
UniProt ID: (Human) P07686, (Mouse) P20060, (Rat) Q6AXR4
Entrez Gene ID: (Human) 3074, (Mouse) 15212, (Rat) 294673
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.