Invitrogen
Manufacturer Code:MA517089
Catalog # PIMA517089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F10 |
Immunogen | Purified recombit fragment of human HEXA expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-hexosaminidase subunit alpha; Beta-N-acetylhexosaminidase subunit alpha; hexosaminidase A (alpha polypeptide); Hexosaminidase subunit A; N-acetyl-beta-glucosaminidase subunit alpha
Gene Aliases: Hex-1; HEXA; TSD
UniProt ID: (Human) P06865, (Mouse) P29416
Entrez Gene ID: (Human) 3073, (Mouse) 15211
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.