Abnova
Manufacturer Code:H00003073M06A
Catalog # 89-104-629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D1 |
Immunogen | HEXA (AAH189271 a.a. &bsim 529 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant HEXA.
For Research Use Only
Protein Aliases: Beta-hexosaminidase subunit alpha; Beta-N-acetylhexosaminidase subunit alpha; hexosaminidase A (alpha polypeptide); Hexosaminidase subunit A; MGC99608; N-acetyl-beta-glucosaminidase subunit alpha; TSD
Gene Aliases: HEXA; TSD
UniProt ID: (Human) P06865
Entrez Gene ID: (Human) 3073
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.