OriGene
Manufacturer Code:TA700484
Catalog # 50-167-7936
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B5 |
Immunogen | Full length human recombit protein of human HES1 produced in HEK293T cell. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHb39; Class B basic helix-loop-helix protein 39; Hairy and enhancer of split 1; Hairy homolog; Hairy-like protein; HES-1 HHL HRY bHLHb39; hHL; Transcription factor HES-1
Gene Aliases: BHLHB39; HES-1; HES1; HHL; HL; HRY
UniProt ID: (Human) Q14469
Entrez Gene ID: (Human) 3280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.