Invitrogen
Manufacturer Code:MA535063
Catalog # PIMA535063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0513 |
Immunogen | A synthesized peptide derived from human Hes1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: BHLHB39 BOS_934 Class B basic helix-loop-helix protein 39 FLJ20408 Hairy and enhancer of split 1 Hairy homolog Hairy-like Hairy-like protein Hes1 Hes-1 hHL Hl HRY RHL Transcription factor HES-1; Hairy and enhancer of split 1; Hairy-like protein; RHL; Transcription factor HES-1
Gene Aliases: bHLHb39; Hes-1; Hes1; Hl; Hry
UniProt ID: (Mouse) P35428, (Rat) Q04666
Entrez Gene ID: (Mouse) 15205, (Rat) 29577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.