Novus Biologicals
Manufacturer Code:NBP267642
Catalog # NBP267642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SC06-21 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: bHLHb39; BHLHB39 bHLHb39hairy homolog (Drosophila) Class B basic helix-loop-helix protein 39 FLJ20408 Hairy and enhancer of split 1 hairy and enhancer of split 1 (Drosophila) Hairy homolog Hairy-like protein Hes1 HES-1 hHL HL HRYHHL transcription factor HES-1; Class B basic helix-loop-helix protein 39; Hairy and enhancer of split 1; Hairy homolog; Hairy-like protein; hHL; Transcription factor HES-1
Gene Aliases: BHLHB39; HES-1; HES1; HHL; HL; HRY
UniProt ID: (Human) Q14469
Entrez Gene ID: (Human) 3280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.