Novus Biologicals
Manufacturer Code:NBP190512
Catalog # NBP190512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:HLRGTHRDSLALFPHMATTAFMQPDHAGIFRVFCATMPHLSRGMGQIYEVSSCDNRDPSEQRYGMIRTFYIAAEEVEWDYAPNKNWEFEKQHVDARGERHGDIFMNRTENWIGSQYKKVVYREYTDGEFVEIKARPPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686F22190 hephaestin-like 1 hephaestin-like protein 1; Ferroxidase HEPHL1; hephaestin-like 1; Hephaestin-like protein 1
Gene Aliases: HEPHL1
UniProt ID: (Human) Q6MZM0
Entrez Gene ID: (Human) 341208
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.