Proteintech
Manufacturer Code:181771AP150UL
Catalog # 50-173-2032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HEPACAM Fusion Protein Ag12870 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GlialCAM HEPACAM Protein hepaCAM; HEPACAM; hepatic and glial cell adhesion molecule; Hepatocyte cell adhesion molecule; Protein hepaCAM
Gene Aliases: 2900042E01Rik; GlialCAM; HEPACAM; Hepn1; MLC2A; MLC2B; RGD1306811
UniProt ID: (Human) Q14CZ8, (Mouse) Q640R3
Entrez Gene ID: (Human) 220296, (Mouse) 72927, (Rat) 300517
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.