Invitrogen
Manufacturer Code:PA5110958
Catalog # PIPA5110958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human HENMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf59 HEN1 HEN1 methyltransferase homolog 1 HENMT1 Small RNA 2&prime-O-methyltransferase; HEN1 methyltransferase homolog 1; hua enhancer 1 homolog 1; Small RNA 2'-O-methyltransferase
Gene Aliases: C1orf59; HEN1; HENMT1
UniProt ID: (Human) Q5T8I9
Entrez Gene ID: (Human) 113802
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.