Novus Biologicals
Manufacturer Code:NBP287559
Catalog # NB035181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human HENMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 1 open reading frame 59 EC 2.1.1.n8 FLJ30525 HEN1 methyltransferase homolog 1 HEN1 methyltransferase homolog 1 (Arabidopsis) HEN1C1orf59hypothetical protein LOC113802 MGC111091; HEN1 methyltransferase homolog 1; hua enhancer 1 homolog 1; Small RNA 2'-O-methyltransferase
Gene Aliases: C1orf59; HEN1; HENMT1
UniProt ID: (Human) Q5T8I9
Entrez Gene ID: (Human) 113802
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.