Invitrogen
Manufacturer Code:PA576833
Catalog # PIPA576833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human HELLS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Helicase lymphoid specific Hells HELLS&lowbarHUMAN LSH Lymphoid specific helicase Lymphoid-specific helicase Nbla10143 PASG Proliferation associated SNF2 like protein Proliferation-associated SNF2-like protein SWI/SNF2 related matrix associated actin dependent regulator of chromatin subfamily A member 6 SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; helicase, lymphoid specific; hells; Lymphocyte-specific helicase; Lymphoid-specific helicase; Proliferation-associated SNF2-like protein; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6; TBC1D12: TBC1 domain family, member 12
Gene Aliases: AI323785; E130115I21Rik; HELLS; ICF4; LSH; Lysh; Nbla10143; PASG; SMARCA6; Tbc1d12; YFK8
UniProt ID: (Human) Q6I7N7, (Mouse) Q60848
Entrez Gene ID: (Human) 3070, (Mouse) 15201, (Rat) 294071
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.