Proteintech
Manufacturer Code:127061AP150UL
Catalog # 50-172-8191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HEBP2 Fusion Protein Ag3387 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf34 C6ORF34B HEBP2 heme binding protein 2 KIAA1244 Placental protein 23 PP23 Protein SOUL SOUL; Heme-binding protein 2; Placental protein 23; PP23; Protein SOUL; putative heme-binding protein
Gene Aliases: C6orf34; C6ORF34B; HEBP2; PP23; SOUL
UniProt ID: (Human) Q9Y5Z4, (Mouse) Q9WU63
Entrez Gene ID: (Human) 23593, (Mouse) 56016, (Rat) 308632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.