Invitrogen
Manufacturer Code:PA523310
Catalog # PIPA523310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (526 KGERIREIRDKFPEV 540&bsim&Hat925 QENGDEAGEGREAKDC 940) of Vigilin/HDLBP protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HBP PRO2900 VGL HDL-binding protein high density lipoprotein-binding protein vigilin; HDL binding protein; HDL-binding protein; high density lipoprotein binding protein (vigilin); High density lipoprotein-binding protein; lipoprotein-binding protein; Vigilin
Gene Aliases: 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDLBP; PRO2900; VGL
UniProt ID: (Human) Q00341, (Mouse) Q8VDJ3
Entrez Gene ID: (Horse) 100057882, (Pig) 100233187, (Dog) 477431, (Human) 3069, (Mouse) 110611, (Rat) 64474
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.