Novus Biologicals
Manufacturer Code:NBP224769SS
Catalog # NBP224769SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 500-550 and 900-950 of human Vigilin was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HBPFLJ16432 HDL-binding protein high density lipoprotein binding protein High density lipoprotein-binding protein PRO2900 VGLvigilin; HDL-binding protein; High density lipoprotein-binding protein; Vigilin
Gene Aliases: HBP; HDLBP; PRO2900; VGL
UniProt ID: (Human) Q00341
Entrez Gene ID: (Human) 3069
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.