Novus Biologicals
Manufacturer Code:NBP303520100UL
Catalog # NB093057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 178-397 of human HDJ2 (NP_001530.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dj-2 DjA1 DnaJ (Hsp40) homolog subfamily A member 1 dnaJ homolog subfamily A member 1 DnaJ protein homolog 2 DNAJ2 hDJ-2 HDJ2 Heat shock 40 kDa protein 4 Heat shock protein J2 heat shock protein DNAJ-like 2 HSDJ HSJ2 HSJ-2 HSPF4hdj-2 Human DnaJ protein 2 NEDD7 neural precursor cell expressed developmentally down-regulated 7; DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ homolog subfamily A member 1; DnaJ protein homolog 2; hDj-2; Heat shock 40 kDa protein 4; Heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ-2; Human DnaJ protein 2; neural precursor cell expressed, developmentally down-regulated 7
Gene Aliases: DJ-2; DjA1; DNAJ2; DNAJA1; hDJ-2; HDJ2; HSDJ; HSJ-2; HSJ2; HSPF4; NEDD7
UniProt ID: (Human) P31689
Entrez Gene ID: (Human) 3301
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.