Novus Biologicals
Manufacturer Code:NBP200487
Catalog # NBP200487
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7A2 |
Immunogen | Full length human recombinant protein of human HDHD1(NP_036212) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5'-PsiMPase; DXF68S1E5'-PsiMPase EC 3.1.3.n6 family with sequence similarity 16 member A X-linked GS1FAM16AX haloacid dehalogenase-like hydrolase domain containing 1 haloacid dehalogenase-like hydrolase domain containing 1A Haloacid dehalogenase-like hydrolase domain-containing protein 1 Haloacid dehalogenase-like hydrolase domain-containing protein 1A HDHD1Apseudouridine-5'-monophosphatase Protein GS1; family with sequence similarity 16, member A, X-linked; haloacid dehalogenase-like hydrolase domain containing 1; haloacid dehalogenase-like hydrolase domain containing 1A; Haloacid dehalogenase-like hydrolase domain-containing protein 1; Haloacid dehalogenase-like hydrolase domain-containing protein 1A; Protein GS1; Pseudouridine-5'-monophosphatase; Pseudouridine-5'-phosphatase
Gene Aliases: DXF68S1E; FAM16AX; GS1; HDHD1; HDHD1A; PUDP
UniProt ID: (Human) Q08623
Entrez Gene ID: (Human) 8226
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.