Proteintech
Manufacturer Code:600641IG150UL
Catalog # 50-173-6218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5A7G3 |
Immunogen | HDGF Fusion Protein Ag1893 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HDGF; HDGF Hepatoma derived growth factor HMG 1L2 HMG1L2; Hepatoma-derived growth factor; High mobility group protein 1-like 2; HMG-1L2
Gene Aliases: AI118077; D3Ertd299e; HDGF; HMG1L2; Tdrm1
UniProt ID: (Human) P51858, (Rat) Q8VHK7, (Mouse) P51859
Entrez Gene ID: (Human) 3068, (Rat) 114499, (Mouse) 15191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.