Invitrogen
Manufacturer Code:PA5113184
Catalog # PIPA5113184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Guanosine-3&prime5&prime-bis(diphosphate) 3&prime-pyrophosphohydrolase MESH1 protein (2-140AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (ppGpp)ase; (ppGpp)ase 1110033O09Rik BOS_20098 C86475 Guanosine-3&prime5&prime-bis(diphosphate) 3&prime-pyrophosphohydrolase MESH1 HD domain containing 3 HD domain-containing protein 3 HDDC3 LOC100339523 MESH1 Metazoan SpoT homolog 1 metazoan SpoT homolog-1 Penta-phosphate guanosine-3&prime-pyrophosphohydrolase RGD1311839; Guanosine-3',5'-bis(diphosphate) 3'-pyrophosphohydrolase MESH1; HD domain-containing protein 3; MESH1; Metazoan SpoT homolog 1; metazoan SpoT homolog-1; Penta-phosphate guanosine-3'-pyrophosphohydrolase
Gene Aliases: (ppGpp)ase; 1110033O09Rik; C86475; HDDC3; MESH1
UniProt ID: (Human) Q8N4P3, (Mouse) Q9D114
Entrez Gene ID: (Human) 374659, (Mouse) 68695
Molecular Function: hydrolase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.