Proteintech
Manufacturer Code:210911AP150UL
Catalog # 50-173-3253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HDDC3 Fusion Protein Ag15342 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (ppGpp)ase; Guanosine-3',5'-bis(diphosphate) 3'-pyrophosphohydrolase MESH1; HD domain containing 3 HD domain containing protein 3 HDDC3; HD domain-containing protein 3; MESH1; Metazoan SpoT homolog 1; metazoan SpoT homolog-1; Penta-phosphate guanosine-3'-pyrophosphohydrolase
Gene Aliases: (ppGpp)ase; 1110033O09Rik; C86475; HDDC3; MESH1; RGD1311839
UniProt ID: (Human) Q8N4P3, (Mouse) Q9D114
Entrez Gene ID: (Human) 374659, (Rat) 308758, (Mouse) 68695
Molecular Function:
hydrolase
pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.