Proteintech
Manufacturer Code:175481AP150UL
Catalog # 50-173-1636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HDAC8 Fusion Protein Ag11696 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HD8; HD8 HDAC8 HDACL1 histone deacetylase 8 RPD3; Histone deacetylase 8; histone deacetylase-like 1
Gene Aliases: 2610007D20Rik; CDA07; CDLS5; HD8; HDAC8; HDACL1; MRXS6; RGD1562895; RPD3; WTS
UniProt ID: (Human) Q9BY41, (Rat) B1WC68, (Mouse) Q8VH37
Entrez Gene ID: (Human) 55869, (Rat) 363481, (Mouse) 70315
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.