OriGene
Manufacturer Code:CF502083
Catalog # 50-166-1021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G7 |
Immunogen | Full length human recombit protein of human HDAC6 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CPBHM HD6 JM21 PPP1R90; HD6; Histone deacetylase 6; protein phosphatase 1, regulatory subunit 90; Tubulin-lysine deacetylase HDAC6
Gene Aliases: CPBHM; HD6; HDAC6; JM21; KIAA0901; PPP1R90
UniProt ID: (Human) Q9UBN7
Entrez Gene ID: (Human) 10013
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.